siRNA Details
Gene Symbol: Parp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181402.3 | NP_852067.1 | 9 | 1053 | ||
GenBank mRNA | AK030559.1 | - | 3 | 430 | ||
AK151393.1 | - | 6 | 743 | |||
BC040269.1 | - | 9 | 1053 | 4035469 |

More Information
Other Species Targeted

Gene Ontology Categories:
