siRNA Details
Gene Symbol: Parp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181402.3 | NP_852067.1 | 7 | 690 | ||
GenBank mRNA | AK030559.1 | - | 1 | 67 | ||
AK039938.1 | - | 3 | 310 | |||
AK133697.1 | - | 7 | 738 | |||
AK151393.1 | - | 4 | 380 | |||
BC040269.1 | - | 7 | 690 | 4035469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
