siRNA Details
Gene Symbol: Tra2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198102.2 | NP_932770.2 | 6 | 887 | ||
GenBank mRNA | AK164994.1 | - | 1 | 28 | ||
AK171708.1 | - | 5 | 2107 | |||
BC023018.1 | - | 3 | 267 | |||
BC057448.1 | - | 6 | 848 | 4016620 | ||
BC058764.1 | - | 7 | 1152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
