siRNA Details
Gene Symbol: Brk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133937.1 | NP_598698.1 | 1, 2 | 132 | ||
GenBank mRNA | AK003342.1 | - | 1, 2 | 147 | ||
AK020128.1 | - | 1, 2 | 147 | |||
AK020277.1 | - | 1, 2 | 148 | |||
AK150689.1 | - | 1, 2 | 147 | |||
AK163236.1 | - | 1, 2 | 147 | |||
AK166585.1 | - | 1, 2 | 135 | |||
AK190298.1 | - | 1, 2 | 71 | |||
AK193342.1 | - | 1, 2 | 71 | |||
BC010582.1 | - | 1, 2 | 132 | 3154777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
