siRNA Details
Gene Symbol: Dyrk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207210.2 | NP_997093.2 | 10, 11 | 1186 | ||
GenBank mRNA | AK077117.1 | - | 10, 11 | 1187 | ||
BC052324.1 | - | 9, 10 | 1240 | 6315342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
