siRNA Details
Gene Symbol: Usb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133954.2 | NP_598715.2 | 3 | 408 | ||
GenBank mRNA | AK135527.1 | - | 3 | 401 | ||
AK160901.1 | - | 3 | 391 | |||
AK168683.1 | - | 3 | 390 | |||
BC016418.1 | - | 3 | 352 | 4224590 |
