siRNA Details
Gene Symbol: Champ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181854.2 | NP_862902.1 | 2 | 2391 | ||
GenBank mRNA | AK129444.1 | - | 2 | 2291 | ||
AK132164.1 | - | 2 | 2354 | |||
AK133990.1 | - | 2 | 2317 | |||
AK170835.1 | - | 2 | 2334 | |||
BC028991.1 | - | 2 | 2303 | 5355869 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
