siRNA Details
Gene Symbol: Champ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181854.2 | NP_862902.1 | 2 | 2108 | ||
GenBank mRNA | AK129444.1 | - | 2 | 2008 | ||
AK132164.1 | - | 2 | 2071 | |||
AK133990.1 | - | 2 | 2034 | |||
AK139963.1 | - | 2 | 2074 | |||
AK170835.1 | - | 2 | 2051 | |||
BC028991.1 | - | 2 | 2020 | 5355869 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
