siRNA Details
Gene Symbol: Enkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198299.1 | NP_938041.1 | 7 | 997 | ||
GenBank mRNA | AK087475.1 | - | 6 | 1160 | ||
BC018168.1 | - | 6 | 1283 | |||
BC023921.1 | - | 6 | 1386 | |||
BC026993.1 | - | 4 | 3064 | |||
BC052498.1 | - | 7 | 997 | 1245862 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
