siRNA Details
Gene Symbol: Snx19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028874.2 | NP_083150.1 | 1 | 587 | ||
GenBank mRNA | AK122231.1 | - | 1 | 808 | ||
AK163810.1 | - | 1 | 816 | |||
BC051164.1 | - | 1 | 808 | |||
BC063262.1 | - | 1 | 587 | 6405897 |
