siRNA Details
Gene Symbol: Rpp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133982.1 | NP_598743.1 | 1 | 517 | ||
GenBank mRNA | AK082822.1 | - | 1 | 587 | ||
AK140177.1 | - | 1 | 594 | |||
BC016085.1 | - | 1 | 517 | 4501535 | ||
BC083064.1 | - | 1 | 577 | 5367203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
