siRNA Details
Gene Symbol: Oaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178644.3 | NP_848759.1 | 4 | 1031 | ||
GenBank mRNA | AK051349.1 | - | 4 | 1031 | ||
AK150279.1 | - | 4 | 1018 | |||
BC025514.1 | - | 4 | 1039 | 5309953 | ||
BC026022.1 | - | 4 | 995 | 5098241 | ||
BC028824.1 | - | 4 | 1066 | 5097873 | ||
BC034836.1 | - | 4 | 997 | 5125217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
