siRNA Details
Gene Symbol: Pwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133993.3 | NP_598754.2 | 6, 7 | 721 | ||
GenBank mRNA | AK009972.1 | - | 6, 7 | 688 | ||
AK011891.1 | - | 6, 7 | 669 | |||
AK075805.1 | - | 6, 7 | 663 | |||
AK082158.1 | - | 6, 7 | 663 | |||
AK149699.1 | - | 6, 7 | 665 | |||
BC003199.1 | - | 6, 7 | 690 | 3586496 | ||
BC023137.1 | - | 6, 7 | 671 | 3986908 |
