siRNA Details
Gene Symbol: Tmem263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013028.2 | NP_001013046.1 | 3 | 571 | ||
GenBank mRNA | AK005698.1 | - | 456 | |||
AK142035.1 | - | 3 | 424 | |||
BC086492.1 | - | 3 | 436 | 30470653 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108739, XM_008765244, XM_006241164, FQ218051, BC166979.
Gene Ontology Categories:
