siRNA Details
Gene Symbol: Cep57l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243074.1 | NP_001230003.1 | 5 | 614 | ||
NM_001243075.1 | NP_001230004.1 | 3 | 305 | |||
NM_026643.5 | NP_080919.3 | 5 | 532 | |||
NM_029132.2 | NP_083408.1 | 5 | 600 | |||
GenBank mRNA | AK010530.1 | - | 5 | 551 | ||
AK012710.1 | - | 3 | 251 | |||
AK015619.1 | - | 4 | 515 | |||
BC021375.1 | - | 5 | 624 | 5038957 | ||
BC138230.1 | - | 5 | 535 | 8861249 | ||
BQ553570.1 | - | 4 | 489 | |||
CD354087.1 | - | 2 | 282 |
