siRNA Details
Gene Symbol: Cep57l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243074.1 | NP_001230003.1 | 8 | 833 | ||
NM_001243075.1 | NP_001230004.1 | 7 | 611 | |||
NM_026643.5 | NP_080919.3 | 8 | 751 | |||
NM_029132.2 | NP_083408.1 | 9 | 906 | |||
GenBank mRNA | AF397908.1 | - | 9 | 834 | ||
AK006744.1 | - | 3 | 137 | |||
AK010530.1 | - | 9 | 857 | |||
AK012710.1 | - | 7 | 557 | |||
BC021375.1 | - | 8 | 843 | 5038957 | ||
BC138230.1 | - | 8 | 754 | 8861249 | ||
CD354087.1 | - | 5 | 501 |
