siRNA Details
Gene Symbol: Zfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034895.3 | NP_001030067.2 | 2, 3 | 326 | ||
GenBank mRNA | AK165530.1 | - | 2, 3 | 314 | ||
BB641594.1 | - | 2, 3 | 326 | |||
BC072571.1 | - | 2, 3 | 302 | |||
CB249573.1 | - | 2, 3 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
