siRNA Details
Gene Symbol: Sec14l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146013.1 | NP_666125.1 | 5 | 427 | ||
GenBank mRNA | AK028954.1 | - | 5 | 388 | ||
BC026948.1 | - | 5 | 427 | 5136664 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
