siRNA Details
Gene Symbol: Tmed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134020.1 | NP_598781.1 | 4 | 449 | ||
GenBank mRNA | AF368921.1 | - | 4 | 480 | ||
AK078425.1 | - | 4 | 460 | |||
AK158876.1 | - | 4 | 482 | |||
AK204643.1 | - | 2 | 144 | |||
BC023041.1 | - | 4 | 449 | 5360236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
