siRNA Details
Gene Symbol: Slc35e4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153142.3 | NP_694782.1 | 2 | 2451 | ||
GenBank mRNA | AK039786.1 | - | 2 | 2452 | ||
BC020181.1 | - | 2 | 1485 | 4488513 | ||
BC095957.1 | - | 2 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
