siRNA Details
Gene Symbol: Tubg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134028.2 | NP_598789.1 | 6 | 732 | ||
GenBank mRNA | AB158481.1 | - | 6 | 526 | ||
BC019652.1 | - | 6 | 712 | 4501003 | ||
BC051439.1 | - | 6 | 594 | 6491052 | ||
BC078446.1 | - | 4 | 1167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
