siRNA Details
Gene Symbol: Rpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133933.4 | NP_598694.3 | 5 | 936 | ||
GenBank mRNA | AK029923.1 | - | 5 | 894 | ||
AK052130.1 | - | 5 | 890 | |||
AK132750.1 | - | 5 | 909 | |||
AK151899.1 | - | 5 | 900 | |||
AK152003.1 | - | 5 | 909 | |||
AK153867.1 | - | 5 | 895 | |||
AK178942.1 | - | 2 | 135 | |||
BC016080.1 | - | 5 | 898 | 4489287 | ||
BC085483.1 | - | 5 | 877 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
