siRNA Details
Gene Symbol: Wdr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014981.1 | NP_001014981.1 | 2, 3 | 392 | ||
GenBank mRNA | AF188123.1 | - | 1, 2 | 154 | ||
AF188124.1 | - | 1, 2 | 154 | |||
AK052730.1 | - | 2, 3 | 393 | |||
AK154451.1 | - | 2, 3 | 372 | |||
BC026618.1 | - | 2, 3 | 311 | |||
BC125573.1 | - | 2, 3 | 260 | 40130088 | ||
BC137785.1 | - | 2, 3 | 260 | 8860804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
