siRNA Details
Gene Symbol: Itga9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133721.2 | NP_598482.2 | 6 | 943 | ||
GenBank mRNA | AJ344342.1 | - | 6 | 946 | ||
AK153585.1 | - | 4 | 405 | |||
BC150824.1 | - | 6 | 849 | 9087735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
