siRNA Details
Gene Symbol: Itga9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133721.2 | NP_598482.2 | 8 | 1079 | ||
GenBank mRNA | AJ344342.1 | - | 8 | 1082 | ||
AK153585.1 | - | 6 | 541 | |||
BC150824.1 | - | 8 | 985 | 9087735 |

More Information
Other Species Targeted

Gene Ontology Categories:
