siRNA Details
Gene Symbol: Gldc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138595.2 | NP_613061.1 | 24 | 3084 | ||
GenBank mRNA | AK010327.1 | - | 4 | 395 | ||
AK080789.1 | - | 24 | 3056 | |||
AK080937.1 | - | 24 | 3074 | |||
BC017135.1 | - | 24 | 3061 | 4511573 |
