siRNA Details
Gene Symbol: Chil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009892.2 | NP_034022.2 | 267 | |||
GenBank mRNA | AK137503.1 | - | 244 | |||
AK154420.1 | - | 242 | ||||
BC061154.1 | - | 242 | 30298306 | |||
BY117835.1 | - | 270 | ||||
D87757.2 | - | 3 | 224 | |||
M94584.2 | - | 236 |
Gene Symbol: Chil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145126.2 | NP_660108.2 | 3 | 239 | ||
GenBank mRNA | AK131914.1 | - | 3 | 240 | ||
AY049765.2 | - | 3 | 224 | |||
AY065557.1 | - | 3 | 224 | |||
BC130015.1 | - | 3 | 228 | 40140436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
