siRNA Details
Gene Symbol: Cdc42ep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026772.2 | NP_081048.1 | 2 | 612 | ||
GenBank mRNA | AK083004.1 | - | 614 | |||
BC034884.1 | - | 1 | 570 | 5257253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
