siRNA Details
Gene Symbol: Gas8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018855.2 | NP_061343.2 | 5 | 642 | ||
GenBank mRNA | AK140135.1 | - | 5 | 621 | ||
AY692443.1 | - | 5 | 641 | |||
BC003779.1 | - | 5 | 557 | 3592695 | ||
DQ189098.1 | - | 4 | 538 | |||
U19859.1 | - | 5 | 663 |

More Information
Other Species Targeted

Gene Ontology Categories:
