siRNA Details
Gene Symbol: Rhox9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023894.1 | NP_076383.1 | 4 | 837 | ||
GenBank mRNA | AF201698.1 | - | 4 | 837 | ||
AK018818.1 | - | 3 | 278 | |||
BC099451.1 | - | 4 | 776 | 30716051 | ||
BC099963.1 | - | 2 | 2377 | 30083999 | ||
DQ058646.1 | - | 4 | 837 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
