siRNA Details
Gene Symbol: Npffr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133192.3 | NP_573455.2 | 2 | 407 | ||
GenBank mRNA | AF236084.1 | - | 1 | 301 | ||
AF330054.1 | - | 1 | 292 | |||
AK050939.1 | - | 2 | 408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
