siRNA Details
Gene Symbol: Mycbpap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170671.2 | NP_733771.2 | 9, 10 | 1171 | ||
GenBank mRNA | AK029929.1 | - | 8, 9 | 1347 | ||
AK166780.1 | - | 2, 3 | 319 | |||
BC027327.1 | - | 5, 6 | 532 | 3991131 |
