siRNA Details
Gene Symbol: E030019B13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053157.1 | - | 5 | 2157 | ||
AK138262.1 | - | 2125 | ||||
BC064451.1 | - | 7 | 1586 | |||
KF856953.1 | - | 8 | 1781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
