siRNA Details
Gene Symbol: Tdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028354.4 | NP_082630.2 | 7, 8 | 939 | ||
GenBank mRNA | AK013427.1 | - | 6, 7 | 414 | ||
AK014855.1 | - | 7, 8 | 919 | |||
AK033868.1 | - | 7, 8 | 937 | |||
AK189362.1 | - | 1, 2 | 56 | |||
BC019804.1 | - | 4, 5 | 263 | 4018534 | ||
BC065162.1 | - | 6, 7 | 909 | 6402921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
