siRNA Details
Gene Symbol: Rab15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB232610.1 | - | 2 | 158 | ||
BC013790.1 | - | 2 | 311 | 2655151 | ||
BC027769.1 | - | 2 | 365 | 5345297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
