siRNA Details
Gene Symbol: Fam84a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029007.2 | NP_083283.2 | 2 | 986 | ||
GenBank mRNA | AK014625.1 | - | 2 | 1002 | ||
BC005488.1 | - | 2 | 986 | 3486254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
