siRNA Details
Gene Symbol: Iars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172015.3 | NP_742012.2 | 25, 26 | 2705 | ||
GenBank mRNA | AK087992.1 | - | 25, 26 | 2692 | ||
AK133856.1 | - | 25, 26 | 2692 | |||
BC059884.1 | - | 10, 11 | 1096 | |||
BC067029.1 | - | 25, 26 | 2692 | 30534067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
