siRNA Details
Gene Symbol: Arrdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042591.1 | NP_001036056.1 | 5 | 1064 | ||
GenBank mRNA | AK084951.1 | - | 5 | 1075 | ||
AK088288.1 | - | 5 | 1079 | |||
AK122499.1 | - | 5 | 1051 | |||
AK163364.1 | - | 5 | 609 | |||
BC023838.1 | - | 2 | 282 | |||
BC031125.1 | - | 3 | 442 | |||
BC044762.1 | - | 5 | 1062 | |||
BC054844.1 | - | 5 | 1064 | 6493441 | ||
BC098474.1 | - | 5 | 1058 | 30094818 | ||
DQ861994.1 | - | 5 | 809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
