siRNA Details
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164743.1 | NP_001158215.1 | 3 | 550 | ||
NM_134069.3 | NP_598830.2 | 4 | 674 | |||
GenBank mRNA | AA986956.1 | - | 2 | 201 | ||
AK128896.1 | - | 4 | 658 | |||
AK143896.1 | - | 4 | 646 | |||
AK165505.1 | - | 4 | 519 | |||
BC018252.1 | - | 4 | 603 | 4240254 | ||
BC024459.1 | - | 4 | 630 | 4975341 | ||
BC034179.1 | - | 4 | 494 | 4972500 | ||
BC034183.1 | - | 4 | 479 | 4973620 |
