siRNA Details
Gene Symbol: Akr1c14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134072.1 | NP_598833.1 | 1 | 114 | ||
GenBank mRNA | AK033537.1 | - | 1 | 132 | ||
AK086887.1 | - | 1 | 134 | |||
AK136528.1 | - | 1 | 131 | |||
AY730283.1 | - | 1 | 76 | |||
BC013482.1 | - | 1 | 114 | 4240772 | ||
BC026628.1 | - | 1 | 113 | 4983237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
