siRNA Details
Gene Symbol: Slain1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198014.2 | NP_932131.2 | 4 | 1284 | ||
GenBank mRNA | AK036190.1 | - | 6 | 1389 | ||
BC025223.1 | - | 2 | 438 | 5010865 | ||
BC079866.1 | - | 4 | 1284 | 6811096 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001014139, BC088867.
Gene Ontology Categories:
