siRNA Details
Gene Symbol: Dock9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081039.1 | NP_001074508.1 | 50 | 5697 | ||
NM_001128307.1 | NP_001121779.1 | 51 | 6000 | |||
NM_001128308.1 | NP_001121780.1 | 50 | 5961 | |||
NM_134074.2 | NP_598835.2 | 49 | 5547 | |||
GenBank mRNA | AK045750.1 | - | 20 | 1979 | ||
AK122431.1 | - | 51 | 5999 | |||
AK134685.1 | - | 3 | 205 | |||
BC009134.1 | - | 8 | 885 | |||
BC046250.1 | - | 44 | 5004 | |||
BC052947.1 | - | 44 | 5016 | |||
BC057692.1 | - | 14 | 1529 | |||
BC157964.1 | - | 49 | 5567 | 9007289 | ||
BC158050.1 | - | 50 | 5892 | 9007354 | ||
BC158127.1 | - | 49 | 5430 |
