siRNA Details
Gene Symbol: Mbnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175341.4 | NP_780550.1 | 5 | 1323 | ||
NM_207515.2 | NP_997398.1 | 5 | 1323 | |||
GenBank mRNA | AK028797.1 | - | 5 | 1288 | ||
AK085709.1 | - | 5 | 1286 | |||
AK151913.1 | - | 5 | 1287 | |||
AK151926.1 | - | 5 | 1287 | |||
AK151957.1 | - | 5 | 1287 | |||
AK153261.1 | - | 5 | 1177 | |||
AK159213.1 | - | 5 | 1281 | |||
AK164372.1 | - | 5 | 1286 | |||
AK167479.1 | - | 5 | 1153 | |||
AK167694.1 | - | 6 | 1088 | |||
AK220470.1 | - | 1297 | ||||
BC020406.1 | - | 4 | 721 | |||
BC023895.1 | - | 5 | 1275 | |||
BC026711.1 | - | 4 | 688 | |||
BC075665.1 | - | 4 | 926 | 30605146 | ||
BC079898.1 | - | 5 | 1272 | |||
GQ906534.1 | - | 4 | 641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
