siRNA Details
Gene Symbol: Thtpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153083.5 | NP_694723.1 | 1 | 801 | ||
GenBank mRNA | AF432864.2 | - | 1 | 632 | ||
AK085173.1 | - | 1 | 496 | |||
AK132479.1 | - | 1 | 493 | |||
BC025562.1 | - | 1 | 785 | 5322971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
