siRNA Details
Gene Symbol: Fam19a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252310.1 | NP_001239239.1 | 2 | 333 | ||
NM_134096.2 | NP_598857.1 | 2 | 285 | |||
GenBank mRNA | AK090194.1 | - | 2 | 159 | ||
AK090307.1 | - | 2 | 335 | |||
AY325124.1 | - | 2 | 286 | |||
BC015306.1 | - | 2 | 286 | 4223181 | ||
BU057253.1 | - | 2 | 289 | |||
BY733542.1 | - | 2 | 335 |

More Information
Other Species Targeted

Gene Ontology Categories:
