siRNA Details
Gene Symbol: Gga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145929.2 | NP_666041.1 | 15 | 2049 | ||
GenBank mRNA | AK080881.1 | - | 15 | 1672 | ||
AK155195.1 | - | 15 | 1689 | |||
AK159771.1 | - | 15 | 1672 | |||
AK170437.1 | - | 15 | 1688 | |||
BC026802.1 | - | 15 | 2049 | 4223300 | ||
BC054422.1 | - | 15 | 1526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
