siRNA Details
Gene Symbol: Qtrtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029128.2 | NP_083404.2 | 8 | 949 | ||
GenBank mRNA | AK015536.1 | - | 1200 | |||
BC017628.1 | - | 9 | 1976 | |||
BC083089.1 | - | 8 | 929 | 30105859 | ||
FM985972.1 | - | 8 | 1042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
