siRNA Details
Gene Symbol: Ildr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285788.1 | NP_001272717.1 | 4 | 570 | ||
NM_001285791.1 | NP_001272720.1 | 4 | 570 | |||
NM_134109.2 | NP_598870.1 | 4 | 570 | |||
GenBank mRNA | AK035504.1 | - | 4 | 499 | ||
AK036844.1 | - | 4 | 496 | |||
AK050806.1 | - | 4 | 516 | |||
AK136284.1 | - | 4 | 570 | |||
BC010485.1 | - | 4 | 510 | 3483341 | ||
BC057644.1 | - | 4 | 567 | 4912789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
