siRNA Details
Gene Symbol: Gpsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134116.5 | NP_598877.1 | 4 | 1058 | ||
GenBank mRNA | AK144833.1 | - | 4 | 1042 | ||
AK146245.1 | - | 4 | 1048 | |||
AK155617.1 | - | 4 | 1059 | |||
AK155863.1 | - | 4 | 736 | |||
BC021942.1 | - | 4 | 648 | 5068831 | ||
BC080804.1 | - | 5 | 761 | 5355970 | ||
BC085186.1 | - | 4 | 1040 | 30680693 |
Gene Symbol: Pbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141763.1 | - | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
