siRNA Details
Gene Symbol: Tecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027179.1 | NP_081455.1 | 6 | 561 | ||
NM_134118.5 | NP_598879.1 | 7 | 606 | |||
GenBank mRNA | AK002384.1 | - | 7 | 576 | ||
AK010513.1 | - | 6 | 497 | |||
AK010984.1 | - | 7 | 580 | |||
AK013558.1 | - | 7 | 559 | |||
AK020708.1 | - | 6 | 555 | |||
AK171718.1 | - | 2 | 313 | |||
AK172506.1 | - | 2 | 291 | |||
BC019984.1 | - | 7 | 592 | 4240045 | ||
BC094046.1 | - | 7 | 706 | 5064002 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
